accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3NOCl

CAS RN 1448636-17-6

IUPAC Name: N-benzyl-N-tert-butyl-4-(chloromethyl)benzamide
Synonyms: N-benzyl-N-tert-butyl-4-(chloromethyl)benzamide
Molecular Weight (g/mol): 315.84
Molecular Formula: C19H22ClNO
InChi Key: AGBSCAUSQCMREP-UHFFFAOYSA-N
SMILES: CC(C)(C)N(CC1=CC=CC=C1)C(=O)C1=CC=C(CCl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results