CAS RN 1449132-02-8
IUPAC Name:
{4-[2-(pyridin-2-yl)-1,3-thiazol-4-yl]phenyl}boronic acid
Synonyms:
4-[2-(pyridin-2-yl)-1,3-thiazol-4-yl]phenylboronic acid
Molecular Weight (g/mol):
282.12
Molecular Formula:
C14H11BN2O2S
InChi Key:
NATNASKGEVWEHX-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC=C(C=C1)C1=CSC(=N1)C1=CC=CC=N1
1
–
1
of
1
results
1
4-[2-(2-Pyridyl)-4-thiazolyl]benzeneboronic acid, 97%, Thermo Scientific™
CAS: 1449132-02-8 Molecular Formula: C14H11BN2O2S Molecular Weight (g/mol): 282.124 MDL Number: MFCD22683028 InChI Key: NATNASKGEVWEHX-UHFFFAOYSA-N Synonym: 4-2-pyridin-2-yl-1,3-thiazol-4-yl phenylboronic acid,4-2-2-pyridyl-4-thiazolyl benzeneboronic acid PubChem CID: 99738175 IUPAC Name: [4-(2-pyridin-2-yl-1,3-thiazol-4-yl)phenyl]boronic acid SMILES: B(C1=CC=C(C=C1)C2=CSC(=N2)C3=CC=CC=N3)(O)O