accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1449132-34-6

IUPAC Name: {5-[(4-chlorophenyl)carbamoyl]-2-fluorophenyl}boronic acid
Synonyms: 5-[(4-chlorophenyl)carbamoyl]-2-fluorophenylboronic acid
Molecular Weight (g/mol): 293.49
Molecular Formula: C13H10BClFNO3
InChi Key: RLWSEMVWIKYUAE-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C=CC(=C1)C(=O)NC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results