CAS RN 1449132-92-6
IUPAC Name:
{4-[2-(pyridin-4-yl)-1,3-thiazol-4-yl]phenyl}boronic acid
Synonyms:
4-[2-(pyridin-4-yl)-1,3-thiazol-4-yl]phenylboronic acid
Molecular Weight (g/mol):
282.12
Molecular Formula:
C14H11BN2O2S
InChi Key:
WPXUKBRXQWYSGF-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC=C(C=C1)C1=CSC(=N1)C1=CC=NC=C1
1
–
1
of
1
results
1
4-[2-(4-Pyridyl)-4-thiazolyl]benzeneboronic acid, 97%, Thermo Scientific™
CAS: 1449132-92-6 Molecular Formula: C14H11BN2O2S Molecular Weight (g/mol): 282.124 MDL Number: MFCD22682954 InChI Key: WPXUKBRXQWYSGF-UHFFFAOYSA-N Synonym: 4-2-pyridin-4-yl-1,3-thiazol-4-yl phenylboronic acid,4-2-4-pyridyl-4-thiazolyl benzeneboronic acid PubChem CID: 99738118 IUPAC Name: [4-(2-pyridin-4-yl-1,3-thiazol-4-yl)phenyl]boronic acid SMILES: B(C1=CC=C(C=C1)C2=CSC(=N2)C3=CC=NC=C3)(O)O