CAS RN 1449135-34-5
IUPAC Name:
{4'-[2-(pyridin-4-yl)-1,3-thiazol-4-yl]-[1,1'-biphenyl]-4-yl}boronic acid
Synonyms:
4'-[2-(pyridin-4-yl)-1,3-thiazol-4-yl]-[1,1'-biphenyl]-4-ylboronic acid
Molecular Weight (g/mol):
358.22
Molecular Formula:
C20H15BN2O2S
InChi Key:
WNUZJIZZWJNXMV-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CSC(=N1)C1=CC=NC=C1
1
–
1
of
1
results
1
4'-[2-(4-Pyridyl)-4-thiazolyl]biphenyl-4-boronic acid, 97%, Thermo Scientific™
CAS: 1449135-34-5 Molecular Formula: C20H15BN2O2S Molecular Weight (g/mol): 358.222 MDL Number: MFCD22682941 InChI Key: WNUZJIZZWJNXMV-UHFFFAOYSA-N Synonym: 4'-2-4-pyridyl-4-thiazolyl biphenyl-4-boronic acid,4'-2-pyridin-4-yl-1,3-thiazol-4-yl-1,1'-biphenyl-4-ylboronic acid PubChem CID: 73996305 IUPAC Name: [4-[4-(2-pyridin-4-yl-1,3-thiazol-4-yl)phenyl]phenyl]boronic acid SMILES: B(C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CSC(=N3)C4=CC=NC=C4)(O)O