accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1449135-55-0

IUPAC Name: {2-fluoro-5-[(3-methylphenyl)carbamoyl]phenyl}boronic acid
Synonyms: 2-fluoro-5-[(3-methylphenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 273.07
Molecular Formula: C14H13BFNO3
InChi Key: DULJUFCUHPACGN-UHFFFAOYSA-N
SMILES: CC1=CC(NC(=O)C2=CC(B(O)O)=C(F)C=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results