accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1449142-61-3

IUPAC Name: (3-{[2-(trifluoromethyl)phenyl]sulfamoyl}phenyl)boronic acid
Synonyms: 3-{[2-(trifluoromethyl)phenyl]sulfamoyl}phenylboronic acid
Molecular Weight (g/mol): 345.1
Molecular Formula: C13H11BF3NO4S
InChi Key: GYJGSTNPUJQMHS-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)S(=O)(=O)NC1=CC=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results