accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3COCH3

CAS RN 14496-35-6

IUPAC Name: dimethyl[(5-methylfuran-2-yl)methyl]amine
Synonyms: dimethyl[(5-methylfuran-2-yl)methyl]amine
Molecular Weight (g/mol): 139.2
Molecular Formula: C8H13NO
InChi Key: IYULRWBZWXMBGW-UHFFFAOYSA-N
SMILES: CN(C)CC1=CC=C(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results