Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,5-Trimethylfurfurylamine 98.0+%, TCI America™
SDP

Catalog No. T25085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T25085G Supplier TCI America Supplier No. T25085G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14496-35-6
Molecular Formula C8H13NO
Molecular Weight (g/mol) 139.20
MDL Number MFCD00014113
InChI Key IYULRWBZWXMBGW-UHFFFAOYSA-N
Synonym 2-(Dimethylaminomethyl)-5-methylfuran
PubChem CID 151918
IUPAC Name dimethyl[(5-methylfuran-2-yl)methyl]amine
SMILES CN(C)CC1=CC=C(C)O1

Specifications

Color Yellow
Boiling Point 164°C
Quantity 5 g
UN Number 2733
Formula Weight 139.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N,5-Trimethylfurfurylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.