accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CIICH3

CAS RN 144981-86-2

IUPAC Name: 2,7-diiodo-9,9-dimethyl-9H-fluorene
Synonyms: 2,7-diiodo-9,9-dimethylfluorene
Molecular Weight (g/mol): 446.07
Molecular Formula: C15H12I2
InChi Key: GYOWFFGLGGCYSQ-UHFFFAOYSA-N
SMILES: CC1(C)C2=C(C=CC(I)=C2)C2=C1C=C(I)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results