Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Diiodo-9,9-dimethylfluorene 98.0+%, TCI America™
SDP

Catalog No. D50325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50325G 5 g
D50321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50325G Supplier TCI America Supplier No. D50325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 144981-86-2
Molecular Formula C15H12I2
Molecular Weight (g/mol) 446.07
MDL Number MFCD12827985
InChI Key GYOWFFGLGGCYSQ-UHFFFAOYSA-N
PubChem CID 12994202
IUPAC Name 2,7-diiodo-9,9-dimethyl-9H-fluorene
SMILES CC1(C)C2=C(C=CC(I)=C2)C2=C1C=C(I)C=C2

Specifications

Melting Point 197°C
Quantity 5 g
Formula Weight 446.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Diiodo-9,9-dimethylfluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.