accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3OOOCH3CH3OO

CAS RN 145-41-5

IUPAC Name: sodium 4-{9a,11a-dimethyl-4,7,11-trioxo-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanoate
Synonyms: sodium 4-{9a,11a-dimethyl-4,7,11-trioxo-dodecahydrocyclopenta[a]phenanthren-1-yl}pentanoate
Molecular Weight (g/mol): 424.51
Molecular Formula: C24H33NaO5
InChi Key: FKJIJBSJQSMPTI-UHFFFAOYNA-M
SMILES: [Na+].CC(CCC([O-])=O)C1CCC2C3C(CC(=O)C12C)C1(C)CCC(=O)CC1CC3=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results