Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Dehydrocholate, TCI America™
SDP

Catalog No. D004325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D004325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D004325G Supplier TCI America Supplier No. D004325G

Chemical Identifiers

CAS 145-41-5
Molecular Formula C24H33NaO5
Molecular Weight (g/mol) 424.51
MDL Number MFCD00066411
InChI Key FKJIJBSJQSMPTI-UHFFFAOYNA-M
Synonym sodiumdehydrocholate
PubChem CID 126969958
IUPAC Name sodium 4-{9a,11a-dimethyl-4,7,11-trioxo-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanoate
SMILES [Na+].CC(CCC([O-])=O)C1CCC2C3C(CC(=O)C12C)C1(C)CCC(=O)CC1CC3=O

Specifications

Quantity 25 g
Formula Weight 424.51
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Dehydrocholate
RTECSNumber FZ2500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.