accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONOOH3COOONNHNNN

CAS RN 145040-37-5

IUPAC Name: 1-{[(cyclohexyloxy)carbonyl]oxy}ethyl 2-ethoxy-1-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}-1H-1,3-benzodiazole-7-carboxylate
Synonyms: 1-{[(cyclohexyloxy)carbonyl]oxy}ethyl 2-ethoxy-3-{[2'-(2H-1,2,3,4-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl]methyl}-1,3-benzodiazole-4-carboxylate
Molecular Weight (g/mol): 610.67
Molecular Formula: C33H34N6O6
InChi Key: GHOSNRCGJFBJIB-UHFFFAOYNA-N
SMILES: CCOC1=NC2=CC=CC(C(=O)OC(C)OC(=O)OC3CCCCC3)=C2N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NNN=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results