accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OOOCH3NCO

CAS RN 145080-94-0

IUPAC Name: methyl 3-(tert-butoxy)-2-isocyanatopropanoate
Synonyms: methyl 3-(tert-butoxy)-2-isocyanatopropanoate
Molecular Weight (g/mol): 201.22
Molecular Formula: C9H15NO4
InChi Key: MMWNVZFZWRLTCL-UHFFFAOYNA-N
SMILES: COC(=O)C(COC(C)(C)C)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results