Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (S)-(+)-2-Isocyanato-3-tert-butoxypropionate 96.0+%, TCI America™
SDP

Catalog No. I04731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I04731G Supplier TCI America Supplier No. I04731G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 145080-94-0
Molecular Formula C9H15NO4
Molecular Weight (g/mol) 201.222
MDL Number MFCD00191536
InChI Key MMWNVZFZWRLTCL-ZETCQYMHSA-N
Synonym (S)-(+)-2-Isocyanato-3-tert-butoxypropionic Acid Methyl Ester
PubChem CID 7019166
IUPAC Name methyl (2S)-2-isocyanato-3-[(2-methylpropan-2-yl)oxy]propanoate
SMILES CC(C)(C)OCC(C(=O)OC)N=C=O

Specifications

Quantity 1 g
Formula Weight 201.22
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (S)-(+)-2-Isocyanato-3-tert-butoxypropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.