accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1451042-18-4

IUPAC Name: N-(2-aminophenyl)-4-{1-[2-(thiophen-3-yl)ethyl]-1H-1,2,3-triazol-4-yl}benzamide
Synonyms: N-(2-aminophenyl)-4-{1-[2-(thiophen-3-yl)ethyl]-1,2,3-triazol-4-yl}benzamide
Molecular Weight (g/mol): 389.48
Molecular Formula: C21H19N5OS
InChi Key: LBLSLSOENGWIHL-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1NC(=O)C1=CC=C(C=C1)C1=CN(CCC2=CSC=C2)N=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results