accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHPH3CCH3H3CCH3

CAS RN 14518-69-5

IUPAC Name: tetrabutylphosphanium hydroxide
Synonyms: tetrabutylphosphonium hydroxide
Molecular Weight (g/mol): 276.45
Molecular Formula: C16H37OP
InChi Key: DFQPZDGUFQJANM-UHFFFAOYSA-M
SMILES: [OH-].CCCC[P+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)