Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentafluorophenylmagnesium Bromide Solution

Catalog No. 442585100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-258-5100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 44-258-5100ML Supplier Sigma Aldrich Supplier No. 442585100MLA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Employed in the synthesis of novel arsonic and arsinic acids as ligands for potential transition-metal ion extractants, and in the synthesis of polyfluoroaryl alkyl sulfides.

Specifications

CAS 14518-69-5
Density 0.989 g/mL at 25°C
Molecular Formula C16H37OP
Linear Formula (CH3CH2CH2CH2)4P(OH)
MDL Number MFCD00068456
Quantity 100 mL
Molecular Weight (g/mol) 276.44 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.