CAS RN 145196-87-8
IUPAC Name:
N'-[2-(4-iodophenyl)ethyl]{[3-(1H-imidazol-5-yl)propyl]sulfanyl}methanimidamide dihydrobromide
Synonyms:
iodophenpropit dihydrobromide
Molecular Weight (g/mol):
576.13
Molecular Formula:
C15H21Br2IN4S
InChi Key:
BOSOGNBLIWPCMS-UHFFFAOYSA-N
SMILES:
Br.Br.NC(SCCCC1=CN=CN1)=NCCC1=CC=C(I)C=C1
1
–
1
of
1
results
1
Tocris Bioscience™ Iodophenpropit dihydrobromide
CAS: 145196-87-8 Molecular Formula: C15H21Br2IN4S