accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14521-80-3

NHNBr

CAS RN 14521-80-3

IUPAC Name: 5-bromo-1H-pyrazole
Synonyms: 3-bromo-2H-pyrazole
Molecular Weight (g/mol): 146.98
Molecular Formula: C3H3BrN2
InChi Key: XHZWFUVEKDDQPF-UHFFFAOYSA-N
SMILES: BrC1=CC=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results