accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 145214-05-7

IUPAC Name: 2-(tributylstannyl)-1,3-oxazole
Synonyms: 2-(tributylstannyl)-1,3-oxazole
Molecular Weight (g/mol): 358.11
Molecular Formula: C15H29NOSn
InChi Key: YOWGRWHKDCHINP-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=NC=CO1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)