Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Tri-N-Butylstannyl)Oxazole

Catalog No. 6386175G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-861-75G 5 g
63-861-71G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 63-861-75G Supplier Sigma Aldrich Supplier No. 6386175G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(Tri-n-butylstannyl)oxazole is a synthetic building block used in Stille coupling reaction.2-(Tri-n-butylstannyl)oxazole can be used as a reactant to prepare: Heteroaromatic compounds via Stille-Migita cross-coupling reaction with (hetero)aryl halides using a palladium catalyst. Ethyl 2-[3-(1,3-oxazol-2-yl)-1H-indazol-1-yl]acetate by reacting with 3-iodoindazole in the presence of Pd(PPh3)4 as a catalyst.

Specifications

CAS 145214-05-7
Density 1.170 g/mL at 25°C, 1.71 g/mL at 25°C
Boiling Point 108°C to 110°C (0.2 mmHg, lit.)
Molecular Formula C15H29NOSn
Linear Formula C15H29NOSn
Refractive Index n20/D 1.4930
MDL Number MFCD06798076
Quantity 5 g
Molecular Weight (g/mol) 358.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.