accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOSN

CAS RN 14527-41-4

IUPAC Name: 1,3-thiazole-5-carboxylic acid
Synonyms: 1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 129.13
Molecular Formula: C4H3NO2S
InChi Key: YZVFSQQHQPPKNX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=CS1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)