Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Thiazolecarboxylic acid, 98%

Catalog No. AC454540050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454540050 5 g
AC454540010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454540050 Supplier Thermo Scientific Chemicals Supplier No. 454540050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 14527-41-4 | C4H3NO2S | 129.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14527-41-4
Molecular Formula C4H3NO2S
Molecular Weight (g/mol) 129.13
MDL Number MFCD03428539
InChI Key YZVFSQQHQPPKNX-UHFFFAOYSA-N
Synonym thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, thiazole-5-carboxylicacid, pubchem14533, acmc-209cuh, 5-thiazole carboxylic acid, thiazole 5-carboxylic acid, ksc177g9t
PubChem CID 84494
IUPAC Name 1,3-thiazole-5-carboxylic acid
SMILES OC(=O)C1=CN=CS1

Specifications

Melting Point 208.0°C to 210.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 129.14
Percent Purity 98%
Chemical Name or Material 5-Thiazolecarboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
EINECSNumber 238-545-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.