CAS RN 14531-55-6
IUPAC Name:
3,5-dimethyl-4-nitro-1H-pyrazole
Synonyms:
3,5-dimethyl-4-nitro-1H-pyrazole
Molecular Weight (g/mol):
141.13
Molecular Formula:
C5H7N3O2
InChi Key:
OFQCJVVJRNPSET-UHFFFAOYSA-N
SMILES:
CC1=C(C(C)=NN1)[N+]([O-])=O
1
–
1
of
1
results
1
3,5-Dimethyl-4-nitropyrazole 98.0+%, TCI America™
CAS: 14531-55-6 Molecular Formula: C5H7N3O2 Molecular Weight (g/mol): 141.13 MDL Number: MFCD00052513 InChI Key: OFQCJVVJRNPSET-UHFFFAOYSA-N PubChem CID: 146172 IUPAC Name: 3,5-dimethyl-4-nitro-1H-pyrazole SMILES: CC1=C(C(=NN1)C)[N+](=O)[O-]