accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OCH3CH3OCH3CH3OGd

CAS RN 14532-05-9

IUPAC Name: gadolinium(3+) tris(propan-2-olate)
Synonyms: gadolinium(3+) tris(propan-2-olate)
Molecular Weight (g/mol): 334.51
Molecular Formula: C9H21GdO3
InChi Key: VJLSFXQJAXVOEQ-UHFFFAOYSA-N
SMILES: [Gd+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results