Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium(III) isopropoxide, 99% (REO), Thermo Scientific Chemicals

Catalog No. AA4137703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4137703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4137703 Supplier Thermo Scientific Chemicals Supplier No. 04137703
Add to Cart

missing translation for 'validMainframeMsg'

Sol-Gel synthesis of gadolinium compounds

Sol-Gel synthesis of gadolinium compounds

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14532-05-9
Molecular Formula C9H21GdO3
Molecular Weight (g/mol) 334.51
MDL Number MFCD00144381
InChI Key VJLSFXQJAXVOEQ-UHFFFAOYSA-N
Synonym gadolinium iii isopropoxide, triisopropoxygadolinium, gadolinium isopropoxide, acmc-20akue, gadolinium triisopropoxide, gadolinium tripropan-2-olate, gadolinium-i-propoxide, gadolinium 3+ ion tris propan-2-olate, gadolinium 3+ tris propan-2-olate
PubChem CID 15047187
IUPAC Name gadolinium(3+);propan-2-olate
SMILES [Gd+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]

Specifications

Color White
Linear Formula Gd[OCH(CH3)2]3
Quantity 1 g
UN Number UN2925
Sensitivity Air and moisture sensitive
Solubility Information Soluble in anhydrous 2-propanol, toluene
Formula Weight 334.51
Percent Purity 99%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Gadolinium(III) isopropoxide
Show More Show Less
Hazard Category H228-H314-H335
Hazard Statement GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Precautionary Statement P210-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.