accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOH3COOH3CIBrOCH3

CAS RN 1453864-75-9

IUPAC Name: [4-(bromomethyl)phenyl](2,4,6-trimethoxyphenyl)iodanium 4-methylbenzene-1-sulfonate
Synonyms: [4-(bromomethyl)phenyl](2,4,6-trimethoxyphenyl)iodanium tosylate
Molecular Weight (g/mol): 635.31
Molecular Formula: C23H24BrIO6S
InChi Key: GXDJDWWDUXFVDQ-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1)S([O-])(=O)=O.COC1=CC(OC)=C([I+]C2=CC=C(CBr)C=C2)C(OC)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results