Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[4-(Bromomethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate 90.0+%, TCI America™
SDP

Catalog No. B5269200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5269200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5269200MG Supplier TCI America Supplier No. B5269200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1453864-75-9
Molecular Formula C23H24BrIO6S
Molecular Weight (g/mol) 635.307
InChI Key GXDJDWWDUXFVDQ-UHFFFAOYSA-M
Synonym [4-(Bromomethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium Tosylate
PubChem CID 71814360
IUPAC Name [4-(bromomethyl)phenyl]-(2,4,6-trimethoxyphenyl)iodanium;4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)[O-].COC1=CC(=C(C(=C1)OC)[I+]C2=CC=C(C=C2)CBr)OC

Specifications

Melting Point 155°C
Quantity 200 mg
Formula Weight 635.31
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material [4-(Bromomethyl)phenyl](2,4,6-trimethoxyphenyl)iodonium p-Toluenesulfonate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.