accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH

CAS RN 1454-85-9

IUPAC Name: heptadecan-1-ol
Synonyms: heptadecyl alcohol
Molecular Weight (g/mol): 256.47
Molecular Formula: C17H36O
InChi Key: GOQYKNQRPGWPLP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCO
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)