accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14543-43-2

IUPAC Name: 3-amino-4-hydroxybenzonitrile
Synonyms: 3-amino-4-hydroxybenzonitrile
Molecular Weight (g/mol): 134.14
Molecular Formula: C7H6N2O
InChi Key: ZEWCASRNRWXXSO-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1O)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results