Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-hydroxybenzonitrile, 97%

Catalog No. AAH3385603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3385603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3385603 Supplier Thermo Scientific Chemicals Supplier No. H3385603

missing translation for 'validMainframeMsg'

CAS: 14543-43-2 | C7H6N2O | 134.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14543-43-2
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.138
MDL Number MFCD00234273
InChI Key ZEWCASRNRWXXSO-UHFFFAOYSA-N
PubChem CID 271258
IUPAC Name 3-amino-4-hydroxybenzonitrile
SMILES C1=CC(=C(C=C1C#N)N)O

Specifications

Melting Point 151°C to 153°C
Quantity 1 g
Sensitivity Air Sensitive
Synonym 2-amino-4-cyanophenol, 2-amino-4-cyano-phenol, benzonitrile, 3-amino-4-hydroxy, 3-amino-4-hydroxybenzenecarbonitrile, 5-cyano-2-hydroxyaniline, acmc-209cv1, 3-amino-4-hydroxy-benzonitrile, benzonitrile,3-amino-4-hydroxy, 5-cyano-2-hydroxy-aniline
Formula Weight 134.14
Percent Purity 97%
Chemical Name or Material 3-Amino-4-hydroxybenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.