accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1455-21-6

IUPAC Name: nonane-1-thiol
Synonyms: 1-nonanethiol
Molecular Weight (g/mol): 160.32
Molecular Formula: C9H20S
InChi Key: ZVEZMVFBMOOHAT-UHFFFAOYSA-N
SMILES: CCCCCCCCCS

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results