Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Nonanethiol

Catalog No. 674273250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-427-3250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 67-427-3250MG Supplier Sigma Aldrich Supplier No. 674273250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Nonanethiol is an alkanethiol, a stabilizer that forms a self-assembled monolayer (SAM) on a variety of substrates. It is used as a thiol based protective coating.1-Nonanethiol is mainly used to form self-organized monolayers on gold, mercury, silver and platinum surfaces. These monolayers functionalize the surface atoms and improve the surface property of the composite.

Specifications

CAS 1455-21-6
Density 0.842 g/mL (at 25°C)
Boiling Point 220°C
Molecular Formula C9H20S
Refractive Index n20/D 1.455 (literature)
Linear Formula CH3(CH2)8 SH
Quantity 250 mg
MDL Number MFCD00004913
Synonym 1-Nonyl mercaptan; Mercaptan C9
Molecular Weight (g/mol) 160.32
Percent Purity 99%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.