accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14552-35-3

IUPAC Name: copper(2+) bis(ethane-1,2-diamine) dihydroxide
Synonyms: copper(2+) bis(ethylenediamine) dihydroxide
Molecular Weight (g/mol): 217.76
Molecular Formula: C4H18CuN4O2
InChi Key: WATCRQGYOIZIHC-UHFFFAOYSA-L
SMILES: [OH-].[OH-].[Cu++].NCCN.NCCN
Quantity 
  • (3)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results