Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(ethylenediamine)copper(II) hydroxide solution

Catalog No. 4423051L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-230-51L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-230-51L Supplier Sigma Aldrich Supplier No. 4423051L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis(ethylenediamine)copper(II) hydroxide solution is a stable complex formed by copper (Cu) ions with ethylenediamine. It is a transition metal solution that can be used as a catalyst for electrochemical reduction process.Copper(II) ethylenediamine complex may be used to evaluate the cation exchange capacity (CEC) of clay materials. It may also be used to modify polyoxoniobate, which is a class of polyoxometalates (POMs) that finds potential application in photocatalytic H2 production.

Specifications

CAS 14552-35-3
Density 1.086 g/mL (at 25°C)
Molecular Formula C4H18CuN4O2
Linear Formula Cu(H2NCH2CH2NH2)2(OH)2
Quantity 1 L
MDL Number MFCD00274628
Synonym Copper(II)-ethylenediame complex
Molecular Weight (g/mol) 217.76
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.