accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHOOOHOHOONOONHOCH3HOHOHO

CAS RN 1456553-26-6

IUPAC Name: N-(2-{[3-acetamido-5-hydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-4-yl]oxy}-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl)acetamide
Synonyms: N-(2-{[3-acetamido-5-hydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-4-yl]oxy}-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl)acetamide
Molecular Weight (g/mol): 545.5
Molecular Formula: C22H31N3O13
InChi Key: HXQAUFSCNOLKJP-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(O)C(O)C(CO)OC1OC1C(O)C(CO)OC(OC2=CC=C(C=C2)[N+]([O-])=O)C1NC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results