Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GalNAc beta(1-3)GlcNAc-beta-pNP 98.0+%, TCI America™
SDP

Catalog No. G03522MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03522MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G03522MG Supplier TCI America Supplier No. G03522MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1456553-26-6
Molecular Formula C22H31N3O13
Molecular Weight (g/mol) 545.50
MDL Number MFCD15072172
InChI Key HXQAUFSCNOLKJP-UHFFFAOYNA-N
Synonym 4-Nitrophenyl 2-Acetamido-2-deoxy-3-O-(2-acetamido-2-deoxy-beta-D-galactopyranosyl)-beta-D-glucopyranoside
PubChem CID 133554204
IUPAC Name N-(2-{[3-acetamido-5-hydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-4-yl]oxy}-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl)acetamide
SMILES CC(=O)NC1C(O)C(O)C(CO)OC1OC1C(O)C(CO)OC(OC2=CC=C(C=C2)[N+]([O-])=O)C1NC(C)=O

Specifications

Color White-Yellow
Quantity 2 mg
Formula Weight 545.50
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material GalNAc beta(1-3)GlcNAc-beta-pNP
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.