CAS RN 1456858-58-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ HG-9-91-01
Potent pan-salt inducible kinases (SIK1, SIK2 and SIK3) inhibitor. Also inhibits Lat-B-induced EP differentiation via nuclear YAP1 accumulation.