CAS RN 1456935-98-0
IUPAC Name:
tris(3,4-difluorophenyl)-1,3,5,2,4,6-trioxatriborinane
Synonyms:
tris(3,4-difluorophenyl)-1,3,5,2,4,6-trioxatriborinane
Molecular Weight (g/mol):
419.69
Molecular Formula:
C18H9B3F6O3
InChi Key:
KIRMDENYWDSJJI-UHFFFAOYSA-N
SMILES:
FC1=C(F)C=C(C=C1)B1OB(OB(O1)C1=CC(F)=C(F)C=C1)C1=CC(F)=C(F)C=C1
1
–
1
of
1
results
1
2,4,6-Tris(3,4-difluorophenyl)boroxin 98.0+%, TCI America™
CAS: 1456935-98-0 Molecular Formula: C18H9B3F6O3 Molecular Weight (g/mol): 419.687 InChI Key: KIRMDENYWDSJJI-UHFFFAOYSA-N Synonym: 3,4-Difluorophenylboronic Anhydride, Tris(3,4-difluorophenyl)cycloboroxane PubChem CID: 20654587 IUPAC Name: 2,4,6-tris(3,4-difluorophenyl)-1,3,5,2,4,6-trioxatriborinane SMILES: B1(OB(OB(O1)C2=CC(=C(C=C2)F)F)C3=CC(=C(C=C3)F)F)C4=CC(=C(C=C4)F)F