Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tris(3,4-difluorophenyl)boroxin 98.0+%, TCI America™
SDP

Catalog No. T24305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24305G Supplier TCI America Supplier No. T24305G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1456935-98-0
Molecular Formula C18H9B3F6O3
Molecular Weight (g/mol) 419.687
InChI Key KIRMDENYWDSJJI-UHFFFAOYSA-N
Synonym 3,4-Difluorophenylboronic Anhydride, Tris(3,4-difluorophenyl)cycloboroxane
PubChem CID 20654587
IUPAC Name 2,4,6-tris(3,4-difluorophenyl)-1,3,5,2,4,6-trioxatriborinane
SMILES B1(OB(OB(O1)C2=CC(=C(C=C2)F)F)C3=CC(=C(C=C3)F)F)C4=CC(=C(C=C4)F)F

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 419.69
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tris(3,4-difluorophenyl)boroxin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.