accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2NOSS

CAS RN 1457-47-2

IUPAC Name: 3-(prop-2-en-1-yl)-2-sulfanylidene-1,3-thiazolidin-4-one
Synonyms: 3-(prop-2-en-1-yl)-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Weight (g/mol): 173.25
Molecular Formula: C6H7NOS2
InChi Key: GYGUTBCTEJBRAN-UHFFFAOYSA-N
SMILES: C=CCN1C(=O)CSC1=S
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)