Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Allylrhodanine 98.0+%, TCI America™
SDP

Catalog No. A14125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14125G 5 g
A141225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14125G Supplier TCI America Supplier No. A14125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1457-47-2
Molecular Formula C6H7NOS2
Molecular Weight (g/mol) 173.248
MDL Number MFCD00044700
InChI Key GYGUTBCTEJBRAN-UHFFFAOYSA-N
Synonym 3-allylrhodanine, rhodanine, 3-allyl, n-allylrhodanine, 3-prop-2-en-1-yl-2-sulfanylidene-1,3-thiazolidin-4-one, allyl-4-oxo-2-thioxothiazolidin, 4-thiazolidinone, 3-2-propenyl-2-thioxo, n-allyl rhodanine, 3-allyl-2-thioxo-thiazolidin-4-one, 3-allyl-2-thioxo-1,3-thiazolidin-4-one, 3-prop-2-en-1-yl-2-thioxo-1,3-thiazolidin-4-one
PubChem CID 73823
IUPAC Name 3-prop-2-enyl-2-sulfanylidene-1,3-thiazolidin-4-one
SMILES C=CCN1C(=O)CSC1=S

Specifications

Melting Point 47°C
Boiling Point 139°C
Quantity 5 g
Formula Weight 173.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Allylrhodanine
EINECSNumber (5)-3405
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.