accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 145783-15-9

H3CSNNClNH2Cl

CAS RN 145783-15-9

IUPAC Name: 4,6-dichloro-2-(propylsulfanyl)pyrimidin-5-amine
Synonyms: 4,6-dichloro-2-(propylsulfanyl)pyrimidin-5-amine
Molecular Weight (g/mol): 238.13
Molecular Formula: C7H9Cl2N3S
InChi Key: CJJLJBFJNXMANZ-UHFFFAOYSA-N
SMILES: CCCSC1=NC(Cl)=C(N)C(Cl)=N1
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)