accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONClNH2NH2N

CAS RN 1458-01-1

IUPAC Name: methyl 3,5-diamino-6-chloropyrazine-2-carboxylate
Synonyms: methyl 3,5-diamino-6-chloropyrazine-2-carboxylate
Molecular Weight (g/mol): 202.6
Molecular Formula: C6H7ClN4O2
InChi Key: KOOBYHRLTYIPTH-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC(Cl)=C(N)N=C1N
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)