Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3,5-Diamino-6-chloropyrazine-2-carboxylate 98.0+%, TCI America™
SDP

Catalog No. M23225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23225G 5 g
M232225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23225G Supplier TCI America Supplier No. M23225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1458-01-1
Molecular Formula C6H7ClN4O2
Molecular Weight (g/mol) 202.60
MDL Number MFCD01928388
InChI Key KOOBYHRLTYIPTH-UHFFFAOYSA-N
Synonym unii-4kat2q9chl, methyl 3,5-diamino-6-chloro-2-pyrazinecarboxylate, 4kat2q9chl, methyl 3, 5-diamino-6-chloropyrazine-2-carboxylate, pyrazinecarboxylic acid, 3,5-diamino-6-chloro-, methyl ester, methyl-3,5-diamino-6-chloropyrazine-2-carboxylate, 3,5-diamino-6-chloropyrazine-2-carboxylic acid methyl ester, pubchem11053, acmc-209cvs, methyl 3,5-diamino-6-chloropyrazinecarboxylate
PubChem CID 73827
IUPAC Name methyl 3,5-diamino-6-chloropyrazine-2-carboxylate
SMILES COC(=O)C1=NC(Cl)=C(N)N=C1N

Specifications

Melting Point 213°C
Quantity 5 g
Formula Weight 202.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,5-Diamino-6-chloropyrazine-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.