accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNHClH2NNHClOHSOHOO

CAS RN 14581-21-6

IUPAC Name: bis((4-chlorophenyl)hydrazine); sulfuric acid
Synonyms: bis(4-chlorophenylhydrazine); sulfuric acid
Molecular Weight (g/mol): 383.24
Molecular Formula: C12H16Cl2N4O4S
InChi Key: TUHGNSPTNVPHDK-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NNC1=CC=C(Cl)C=C1.NNC1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results