accessibility menu, dialog, popup

UserName

Viewing profile as user:
OPPPdOOOCH3CH3

CAS RN 14588-08-0

IUPAC Name: (acetyloxy)palladiobis(ylium) acetate; bis(triphenylphosphane)
Synonyms: (acetyloxy)palladiobis(ylium) acetate; bis(triphenylphosphine)
Molecular Weight (g/mol): 749.09
Molecular Formula: C40H36O4P2Pd
InChi Key: YOUIUHDWQIUKAO-UHFFFAOYSA-L
SMILES: CC(=O)O[Pd++]OC(C)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)