Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diacetatobis(triphenylphosphine)palladium(II), Pd 14.2%

Catalog No. AAH6185309
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6185309 10 g
AAH6185303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6185309 Supplier Thermo Scientific Chemicals Supplier No. H6185309

missing translation for 'validMainframeMsg'

CAS: 14588-08-0 | C40H36O4P2Pd | 749.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14588-08-0
Molecular Formula C40H36O4P2Pd
Molecular Weight (g/mol) 749.09
MDL Number MFCD00010013
InChI Key YOUIUHDWQIUKAO-UHFFFAOYSA-L
Synonym bis triphenylphosphinepalladium acetate
PubChem CID 73357379
IUPAC Name (acetyloxy)palladiobis(ylium) acetate; bis(triphenylphosphane)
SMILES CC(=O)O[Pd++]OC(C)=O.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 136°C
Linear Formula Pd[P(C6H5)3]2·2CH3CO2
Quantity 10 g
Formula Weight 749.09
Concentration or Composition (by Analyte or Components) Pd 14.2%
Physical Form Solid
Chemical Name or Material Diacetatobis(triphenylphosphine)palladium(II)
EINECSNumber 238-628-4
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.